Learn More
Dimethyl naphthalene-2,6-dicarboxylate, 99+%
Description
Dimethyl naphthalene-2,6-dicarboxylate is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dimethyl naphthalene-2,6-dicarboxylate is used as a pharmaceutical intermediate.
Solubility
Soluble in hot toluene (very faint turbidity). Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 187°C to 193°C |
| Flash Point | 232°C (449°F) |
| Quantity | 25 g |
| Beilstein | 987259 |
| Solubility Information | Soluble in hot toluene (very faint turbidity). Insoluble in water. |
| Formula Weight | 244.25 |
| Percent Purity | ≥99% |
| Chemical Name or Material | Dimethyl naphthalene-2,6-dicarboxylate |
Safety and Handling
| EINECSNumber | 212-661-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.