Learn More
Sigma Organic Chemistry Dimethyl phthalate | 4L | 131-11-3 | MFCD00008425 | >=99%
Supplier: Sigma Organic Chemistry 5250814L

Dimethyl phthalate, 4L
About This Item:
Synonym(s): DMP
Linear Formula: C6H4-1,2-(CO2CH3)2
Molecular Weight: 194.18
CAS Number: 131-11-3
MDL Number: MFCD00008425
UNSPSC Code: 12162002
Purity: >=99%
Boiling Point: 282 C (lit.)
Melting Point: 2 C (lit.)
Density: 1.19 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.515 (lit.)
Storage: room temp
EINECS Number: 205-011-6
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.