Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfide Borane 90.0+%, TCI America™
SDP

Catalog No. D1843100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1843100ML 100 mL
D184325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1843100ML Supplier TCI America Supplier No. D1843100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 13292-87-0
Molecular Formula C2H9BS
Molecular Weight (g/mol) 75.96
MDL Number MFCD00013189
InChI Key RMHDLBZYPISZOI-UHFFFAOYSA-N
Synonym dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms
PubChem CID 9833925
IUPAC Name (methylsulfanyl)methane borane
SMILES B.CSC

Specifications

Color Yellow
Quantity 100 mL
UN Number 3399
Formula Weight 75.96
Percent Purity ≥90.0% (T)
Physical Form Liquid
Chemical Name or Material Dimethyl Sulfide Borane
RTECSNumber PV5080000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.