Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide, ACS Reagent (DMSO), Reagents

Catalog No. 01338003
Change view
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Glass Bottle
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-338-003 4 L Glass Bottle
01-338-004 4 L Poly Bottle
01-338-002 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-338-003 Supplier Reagents Holdings Llc Supplier No. C11966504C
Add to Cart
Add to Cart

Dimethyl Sulfoxide, ACS Reagent (DMSO), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym DMSO, Methylsulfinylmethane
IUPAC Name methanesulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.5°C
Color Colorless
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 1.101
Boiling Point 189°C
Grade ACS
Chemical Name or Material Dimethyl Sulfoxide, ACS Reagent (DMSO)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.