Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide, Anhydrous, septum-sealed, >99.8%, Solstice

Catalog No. 6003014 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
6 x 100 mL
6 x 1 L
Packaging:
Septum-sealed Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
600-30-14 6 x 100 mL Septum-sealed Bottle
600-30-15 6 x 1 L Septum-sealed Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 600-30-14 Supplier Solstice Supplier No. AS0810100

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methanesulfinylmethane
SMILES CS(C)=O

Specifications

CAS Max % 100.0000%
Quantity 6 x 100 mL
Packaging Septum-sealed Bottle
Physical Form anhydrous
Water 50 ppm max.
Beilstein 506008
Solubility Information Miscible in Water at 20°C
Percent Purity >99.8%
Chemical Name or Material Dimethyl Sulfoxide, Anhydrous, septum-sealed, for applications requiring low water content, >99.8%
ShelfLife 365 days from date of manufacture
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.