Learn More
Specifications
Specifications
| Melting Point | 18.4°C |
| Acidity | Pass Test (USP) |
| Appearance | Clear, Colorless Liquid |
| Decomposition Information | >190°C |
| Density | 1.100g/cm³ |
| Quantity | 500 mL |
| Flash Point | 87°C (188.6°F) |
| Identification | Pass Test (USP, EP/BP) |
| Packaging | HDPE Bottle |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Combustible liquid. DMSO readily penetrates skin and may carry other dissolved chemicals into the body. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. NFPA Health:2 Flammability:2 Instability:0 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.