Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl sulfoxide-d{6}, 99.5% (Isotopic)

Catalog No. AAA1689318
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1689318 50 g
AAA1689309 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1689318 Supplier Thermo Scientific Chemicals Supplier No. A1689318
  • $271.00
    $203.65 / Each of 1

    Save $67.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2206-27-1 | C2H6OS | 84.17 g/mol

DMSO-d6 is a solvent of choice in NMR spectroscopy, owing to its unique characteristics of dissolving both nonpolar and polar compounds, miscibility with almost all organic solvents and permitting high temperature dynamic NMR studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DMSO-d6 is a solvent of choice in NMR spectroscopy, owing to its unique characteristics of dissolving both nonpolar and polar compounds, miscibility with almost all organic solvents and permitting high temperature dynamic NMR studies.

Solubility
Miscible with water and with almost all organic solvents such as alcohols, esters, ketones, chlorinated solvents and aromatic hydrocarbons.

Notes
NMR solvent. Dimethyl Sulfoxide (DMSO) has a melting point of 20°C, freezing close to room temperature. Sometimes when it is delivered it will be in a solid state. To return the material to a liquid state, thaw it in a warm water bath. Care must be taken to prevent water contamination. It is stable, combustible and moisture sensitive. Incompatible with acid chlorides, strong acids, phosphorus halides, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2206-27-1
Molecular Formula C2H6OS
Molecular Weight (g/mol) 84.17
MDL Number MFCD00002090
InChI Key IAZDPXIOMUYVGZ-WFGJKAKNSA-N
Synonym dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide
PubChem CID 75151
IUPAC Name trideuterio(trideuteriomethylsulfinyl)methane
SMILES [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]

Specifications

Quantity 50 g
Sensitivity Hygroscopic
Refractive Index 1.476
Flash Point 88°C (190°F)
Density 1.18 g/mL
Melting Point 20°C
Boiling Point 190°C
Assay Percent Range 99.5%
Solubility Information Miscible with water and with almost all organic solvents such as alcohols,esters,ketones,chlorinated solvents and aromatic hydrocarbons.
Formula Weight 84.17
Chemical Name or Material Dimethyl Sulfoxide-d6
Show More Show Less
Hazard Category H227
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 218-617-0
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.