CAS RN 2206-27-1
Methyl sulfoxide-d6, for NMR, packaged in 0.75 ml ampoules, 99.9 atom % D
CAS: 2206-27-1 Molecular Formula: C2H6OS Molecular Weight (g/mol): 84.17 MDL Number: MFCD00002090 InChI Key: IAZDPXIOMUYVGZ-WFGJKAKNSA-N Synonym: dimethyl sulfoxide-d6,dmso-d6,dimethylsulfoxide-d6,deuterated dmso,methane-d3,sulfinylbis-9ci,methane-d3, sulfinylbis,hexadeuterodimethyl sulfoxide,methane-d3-, sulfinylbis,methanesulfinylmethyl hydrogen,dimethyl-d6 sulfoxide PubChem CID: 75151 IUPAC Name: trideuterio(trideuteriomethylsulfinyl)methane SMILES: [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
Methyl sulfoxide-d6, for NMR, 99.5+ atom % D
CAS: 2206-27-1 Molecular Formula: C2H6OS Molecular Weight (g/mol): 84.17 MDL Number: MFCD00002090 InChI Key: IAZDPXIOMUYVGZ-WFGJKAKNSA-N Synonym: dimethyl sulfoxide-d6,dmso-d6,dimethylsulfoxide-d6,deuterated dmso,methane-d3,sulfinylbis-9ci,methane-d3, sulfinylbis,hexadeuterodimethyl sulfoxide,methane-d3-, sulfinylbis,methanesulfinylmethyl hydrogen,dimethyl-d6 sulfoxide PubChem CID: 75151 SMILES: [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
Methyl sulfoxide-d6, for NMR, 99+ atom % D
CAS: 2206-27-1 Molecular Formula: C2H6OS Molecular Weight (g/mol): 84.17 MDL Number: MFCD00002090 InChI Key: IAZDPXIOMUYVGZ-WFGJKAKNSA-N Synonym: dimethyl sulfoxide-d6,dmso-d6,dimethylsulfoxide-d6,deuterated dmso,methane-d3,sulfinylbis-9ci,methane-d3, sulfinylbis,hexadeuterodimethyl sulfoxide,methane-d3-, sulfinylbis,methanesulfinylmethyl hydrogen,dimethyl-d6 sulfoxide PubChem CID: 75151 IUPAC Name: trideuterio(trideuteriomethylsulfinyl)methane SMILES: [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
Methyl sulfoxide-d6, for NMR, 99.9 atom % D
CAS: 2206-27-1 Molecular Formula: C2H6OS Molecular Weight (g/mol): 84.17 MDL Number: MFCD00002090 InChI Key: IAZDPXIOMUYVGZ-WFGJKAKNSA-N Synonym: dimethyl sulfoxide-d6,dmso-d6,dimethylsulfoxide-d6,deuterated dmso,methane-d3,sulfinylbis-9ci,methane-d3, sulfinylbis,hexadeuterodimethyl sulfoxide,methane-d3-, sulfinylbis,methanesulfinylmethyl hydrogen,dimethyl-d6 sulfoxide PubChem CID: 75151 SMILES: [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
Methyl sulfoxide-d6, for NMR, with 0.03% TMS, 99.9 atom% D
CAS: 2206-27-1 Molecular Formula: C2H6OS Molecular Weight (g/mol): 84.17 MDL Number: MFCD00002090 InChI Key: IAZDPXIOMUYVGZ-WFGJKAKNSA-N Synonym: dimethyl sulfoxide-d6,dmso-d6,dimethylsulfoxide-d6,deuterated dmso,methane-d3,sulfinylbis-9ci,methane-d3, sulfinylbis,hexadeuterodimethyl sulfoxide,methane-d3-, sulfinylbis,methanesulfinylmethyl hydrogen,dimethyl-d6 sulfoxide PubChem CID: 75151 IUPAC Name: trideuterio(trideuteriomethylsulfinyl)methane SMILES: [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
Methyl sulfoxide-d6, for NMR, packaged in 1.00 ml ampoules, 99.9 atom % D
CAS: 2206-27-1 Molecular Formula: C2H6OS Molecular Weight (g/mol): 84.17 MDL Number: MFCD00002090 InChI Key: IAZDPXIOMUYVGZ-WFGJKAKNSA-N Synonym: dimethyl sulfoxide-d6,dmso-d6,dimethylsulfoxide-d6,deuterated dmso,methane-d3,sulfinylbis-9ci,methane-d3, sulfinylbis,hexadeuterodimethyl sulfoxide,methane-d3-, sulfinylbis,methanesulfinylmethyl hydrogen,dimethyl-d6 sulfoxide PubChem CID: 75151 IUPAC Name: trideuterio(trideuteriomethylsulfinyl)methane SMILES: [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
Dimethyl sulfoxide-D6 deuteration degree min 99.8% for NMR spectroscopy MagniSolv™, MilliporeSigma™
CAS: 2206-27-1 Molecular Formula: C2H6OS Molecular Weight (g/mol): 84.17 InChI Key: IAZDPXIOMUYVGZ-WFGJKAKNSA-N Synonym: Hexadeuterodimethyl sulfoxide,DMSO deuterated IUPAC Name: (²H₃)methanesulfinyl(²H₃)methane SMILES: [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
Dimethyl sulfoxide-d{6}, 99.5% (Isotopic)
CAS: 2206-27-1 Molecular Formula: C2H6OS Molecular Weight (g/mol): 84.17 MDL Number: MFCD00002090 InChI Key: IAZDPXIOMUYVGZ-WFGJKAKNSA-N Synonym: dimethyl sulfoxide-d6,dmso-d6,dimethylsulfoxide-d6,deuterated dmso,methane-d3,sulfinylbis-9ci,methane-d3, sulfinylbis,hexadeuterodimethyl sulfoxide,methane-d3-, sulfinylbis,methanesulfinylmethyl hydrogen,dimethyl-d6 sulfoxide PubChem CID: 75151 IUPAC Name: trideuterio(trideuteriomethylsulfinyl)methane SMILES: [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
Dimethyl sulfoxide - D6, deuteration degree min. 99.8% for NMR spectroscopy, MagniSolv™, MilliporeSigma™
CAS: 2206-27-1 Molecular Formula: C2H6OS Molecular Weight (g/mol): 84.17 MDL Number: MFCD00002090 InChI Key: IAZDPXIOMUYVGZ-WFGJKAKNSA-N Synonym: dimethyl sulfoxide-d6,dmso-d6,dimethylsulfoxide-d6,deuterated dmso,methane-d3,sulfinylbis-9ci,methane-d3, sulfinylbis,hexadeuterodimethyl sulfoxide,methane-d3-, sulfinylbis,methanesulfinylmethyl hydrogen,dimethyl-d6 sulfoxide PubChem CID: 75151 IUPAC Name: (²H₃)methanesulfinyl(²H₃)methane SMILES: [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
Dimethyl sulfoxide-d{6}, 99.9% (Isotopic)
CAS: 2206-27-1 Molecular Formula: C2H6OS Molecular Weight (g/mol): 84.17 MDL Number: MFCD00002090 InChI Key: IAZDPXIOMUYVGZ-WFGJKAKNSA-N Synonym: dimethyl sulfoxide-d6,dmso-d6,dimethylsulfoxide-d6,deuterated dmso,methane-d3,sulfinylbis-9ci,methane-d3, sulfinylbis,hexadeuterodimethyl sulfoxide,methane-d3-, sulfinylbis,methanesulfinylmethyl hydrogen,dimethyl-d6 sulfoxide PubChem CID: 75151 SMILES: [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
Methyl sulfoxide-d6, for NMR, 99.8 atom % D
CAS: 2206-27-1 Molecular Formula: C2H6OS Molecular Weight (g/mol): 84.17 MDL Number: MFCD00002090 InChI Key: IAZDPXIOMUYVGZ-WFGJKAKNSA-N Synonym: dmso-d6,dimethyl-d6 sulfoxide,methanesulfinylmethyl hydrogen,dimethylsulfoxide-d6,hexadeuterodimethyl sulfoxide,methane-d3, sulfinylbis,methane-d3,sulfinylbis-9ci,deuterated dmso,methane-d3-, sulfinylbis,dimethyl sulfoxide-d6 PubChem CID: 75151 SMILES: [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
Dimethyl sulfoxide - D6, w/TMS(0.03vol.%), deuteratn. deg. min. 99.8% for NMR MagniSolv™, MilliporeSigma™
CAS: 2206-27-1 Molecular Formula: C2H6OS Molecular Weight (g/mol): 84.17 MDL Number: MFCD00002090 InChI Key: IAZDPXIOMUYVGZ-WFGJKAKNSA-N Synonym: dimethyl sulfoxide-d6,dmso-d6,dimethylsulfoxide-d6,deuterated dmso,methane-d3,sulfinylbis-9ci,methane-d3, sulfinylbis,hexadeuterodimethyl sulfoxide,methane-d3-, sulfinylbis,methanesulfinylmethyl hydrogen,dimethyl-d6 sulfoxide PubChem CID: 75151 IUPAC Name: (²H₃)methanesulfinyl(²H₃)methane SMILES: [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
Methyl sulfoxide-d6, for NMR, packaged in 0.50 ml ampoules, 99.9 atom % D
CAS: 2206-27-1 Molecular Formula: C2H6OS Molecular Weight (g/mol): 84.17 MDL Number: MFCD00002090 InChI Key: IAZDPXIOMUYVGZ-WFGJKAKNSA-N Synonym: dimethyl sulfoxide-d6,dmso-d6,dimethylsulfoxide-d6,deuterated dmso,methane-d3,sulfinylbis-9ci,methane-d3, sulfinylbis,hexadeuterodimethyl sulfoxide,methane-d3-, sulfinylbis,methanesulfinylmethyl hydrogen,dimethyl-d6 sulfoxide PubChem CID: 75151 IUPAC Name: trideuterio(trideuteriomethylsulfinyl)methane SMILES: [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
Methyl sulfoxide-d6, for NMR, contains 0.03% TMS, 99.8 atom% D
CAS: 2206-27-1 Molecular Formula: C2H6OS Molecular Weight (g/mol): 84.17 MDL Number: MFCD00002090 InChI Key: IAZDPXIOMUYVGZ-WFGJKAKNSA-N Synonym: dmso-d6,dimethyl sulfoxide-d6,deuterated dmso,dimethylsulfoxide-d6,methane-d3,sulfinylbis-9ci,methane-d3-, sulfinylbis,hexadeuterodimethyl sulfoxide,methanesulfinylmethyl hydrogen,dimethyl-d6 sulfoxide,methane-d3, sulfinylbis PubChem CID: 75151 SMILES: [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]
Methyl sulfoxide-d6, for NMR, 100.0 atom % D, min. 99.96 atom % D
- 1
- 2