Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl sulfoxide-d{6}, 99.9% (Isotopic)

Catalog No. aa3651746
Change view
Click to view available options
Quantity:
5 x 1 g
10 x 1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3651746 5 x 1 g
AA3651748 10 x 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3651746 Supplier Thermo Scientific Chemicals Supplier No. 03651746
Only null left
Add to Cart

CAS: 2206-27-1 | C2H6OS | 84.17 g/mol

Deuterated DMSO is a common solvent used in NMR spectroscopy. It is in combination with pyridine-d5 plays an important role for solution-state 2D NMR of ball-milled plant cell wall gel. Low-energy collisions of ionized acetone-d6, DMSO-d6 and pyridine-d5 with alkanethiolate is used to prepare self-assembled monolayer surfaces of gold.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2206-27-1
Molecular Formula C2H6OS
Molecular Weight (g/mol) 84.17
MDL Number MFCD00002090
InChI Key IAZDPXIOMUYVGZ-WFGJKAKNSA-N
Synonym dimethyl sulfoxide-d6, dmso-d6, dimethylsulfoxide-d6, deuterated dmso, methane-d3,sulfinylbis-9ci, methane-d3, sulfinylbis, hexadeuterodimethyl sulfoxide, methane-d3-, sulfinylbis, methanesulfinylmethyl hydrogen, dimethyl-d6 sulfoxide
PubChem CID 75151
SMILES [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]

Specifications

Physical Form Liquid
Quantity 5 x 1 g
Sensitivity Hygroscopic
Refractive Index 1.476
Density 1.18 g/mL
Flash Point 88°(190°F)
Melting Point 20°C
Boiling Point 190°C
Assay Percent Range 99.9% (Isotopic)
Solubility Information Miscible with water and diethyl ether.
IUPAC Name trideuterio(trideuteriomethylsulfinyl)methane
Formula Weight 84.17
Percent Purity 99.9%
Chemical Name or Material Dimethyl Sulfoxide, Isotopic
Show More Show Less
Hazard Category H227
Hazard Statement GHS H Statement
H227
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 218-617-0
TSCA No
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.