Learn More
Cambridge Isotope Laboratories Dimethyl Sulfoxide-d6, 2206-27-1, MFCD00002090, 25 g
Supplier:  Cambridge Isotope Laboratories DLM1025
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Chemical Formula: CD3SOCD3, MW: 84.17, 99.9%, Synonyms: DMSO-D6; Methyl sulfoxide-D6; Hexadeuterodimethyl sulfoxide
Catalog No. NC9159575
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.