Learn More
Dimethyl sulfoxide - D6, deuteration degree min. 99.8% for NMR spectroscopy, MagniSolv™, MilliporeSigma™
Shop All MilliporeSigma ProductsSpecifications
Specifications
| Physical Form | Liquid |
| Packaging | Glass Ampule |
| Color | Colorless |
| Quantity | 10 x 0.5 mL |
| Vapor Pressure | 2.5 hPa at 20°C (68°F) |
| Odor | Characteristic |
| Assay Percent Range | 99.8% min. |
| Formula Weight | 84.17 |
| Chemical Name or Material | Dimethyl sulfoxide - D6 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.