Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl sulfoxide-D6 deuteration degree min 99.8% for NMR spectroscopy MagniSolv™, MilliporeSigma™

Catalog No. M1034240009 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 x 0.75 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1034240009 10 x 0.75 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1034240009 Supplier MilliporeSigma Supplier No. 1.03424.0009
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2206-27-1
Molecular Formula C2H6OS
Molecular Weight (g/mol) 84.17
InChI Key IAZDPXIOMUYVGZ-WFGJKAKNSA-N
Synonym Hexadeuterodimethyl sulfoxide, DMSO deuterated
IUPAC Name (²H₃)methanesulfinyl(²H₃)methane
SMILES [2H]C([2H])([2H])S(=O)C([2H])([2H])[2H]

Specifications

Packaging Glass Ampule
Density 1.19 g/cm3 (at 20°C)
Vapor Pressure 2.5 hPa (20°C)
Flash Point 88°C
Quantity 10 x 0.75 mL
Melting Point 20.2°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.