Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl trans-stilbene-4,4'-dicarboxylate, 98+%

Catalog No. AAL1196903
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1196903 1 g
AAL11969MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1196903 Supplier Thermo Scientific Chemicals Supplier No. L1196903
Add to Cart

CAS: 34541-73-6 | C18H16O4 | 296.322 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34541-73-6
Molecular Formula C18H16O4
Molecular Weight (g/mol) 296.322
MDL Number MFCD00039527
InChI Key JOODVYOWCWQPMV-ONEGZZNKSA-N
Synonym e-dimethyl 4,4'-ethene-1,2-diyl dibenzoate, dimethyl 4,4'-stilbenedicarboxylate, dimethyl trans-stilbene-4,4'-dicarboxylate, dimethyl stilbene-4,4'-dicarboxylate, benzoic acid, 4,4'-1e-1,2-ethenediylbis-, dimethyl ester, 4,4'-bis carbomethoxy stilbene, methyl 4-1e-2-4-methoxycarbonyl phenyl vinyl benzoate, methyl 4-e-2-4-methoxycarbonyl phenyl ethenyl benzoate, dimethyl 4,4/'-stilbenedicarboxylate
PubChem CID 5378468
IUPAC Name methyl 4-[(E)-2-(4-methoxycarbonylphenyl)ethenyl]benzoate
SMILES COC(=O)C1=CC=C(C=C1)C=CC2=CC=C(C=C2)C(=O)OC

Specifications

Melting Point 234°C to 236°C
Quantity 1 g
Beilstein 2471189
Formula Weight 296.33
Percent Purity ≥98%
Chemical Name or Material Dimethyl trans-stilbene-4,4'-dicarboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.