Learn More
Apexbio Technology LLC Anti-CD38 Antibody (Daratumumab) 100ug
SDP
Supplier:  Apexbio Technology LLC F13700.1
Encompass_Preferred
Daratumumab (CAS 945721-28-8) is a monoclonal antibody targeting CD38 a surface glycoprotein highly expressed on plasma cells especially in multiple myeloma (MM) cells It is designed to selectively bind CD38 thereby mediating tumor cell lysis via multiple immune effector mechanisms Daratumumab exerts its biological activity primarily through antibody-dependent cell-mediated cytotoxicity (ADCC) complement-dependent cytotoxicity (CDC) antibody-dependent cellular phagocytosis (ADCP) and direct induction of apoptosis Additionally it inhibits adhesion of MM cells to bone marrow stromal cells potentially increasing sensitivity to proteasome inhibitors Based on these pharmacological properties Daratumumab holds research potential in therapeutic strategies targeting multiple myeloma
Catalog No. 50-000-91828
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.