Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylamine Borane 95.0+%, TCI America™
SDP

Catalog No. D1842100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1842100G 100 g
D184225G 25 g
D1842500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D1842100G Supplier TCI America Supplier No. D1842100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 74-94-2
Molecular Formula C2H10BN
Molecular Weight (g/mol) 58.92
MDL Number MFCD00051068
InChI Key FTDUHBOCJSQEKS-UHFFFAOYSA-N
Synonym Borane - Dimethylamine Complex, DMAB
IUPAC Name dimethylamine borane
SMILES B.CNC

Specifications

Melting Point 37°C
Color White
Quantity 100 g
UN Number 2926
Formula Weight 58.92
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethylamine Borane
EINECSNumber (2)-2102
RTECSNumber IP9450000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.