Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylammonium Dimethyldithiocarbamate 95.0+%, TCI America™
SDP

Catalog No. D071525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D071525G 25 g
D07155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D071525G Supplier TCI America Supplier No. D071525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 598-64-1
Molecular Formula C5H14N2S2
Molecular Weight (g/mol) 166.301
MDL Number MFCD00067267
InChI Key UVOFGKIRTCCNKG-UHFFFAOYSA-N
Synonym Dimethyldithiocarbamic Acid Dimethylammonium Salt
PubChem CID 11728
IUPAC Name dimethylazanium;N,N-dimethylcarbamodithioate
SMILES C[NH2+]C.CN(C)C(=S)[S-]

Specifications

Melting Point 132°C
Color White-Yellow
Quantity 25 g
Formula Weight 166.30
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethylammonium Dimethyldithiocarbamate
EINECSNumber (2)-2893
RTECSNumber FA0250000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.