Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyldi-n-octadecylammonium bromide, 99%, Thermo Scientific Chemicals

Catalog No. AAA1824403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1824403 1 g
AAA1824406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1824403 Supplier Thermo Scientific Chemicals Supplier No. A1824403

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3700-67-2
Molecular Formula C38H80BrN
Molecular Weight (g/mol) 630.969
MDL Number MFCD00041975
InChI Key PSLWZOIUBRXAQW-UHFFFAOYSA-M
Synonym dimethyldioctadecylammonium bromide, n,n-dimethyl-n-octadecyloctadecan-1-aminium bromide, dimethyldistearylammonium bromide, 1-octadecanaminium, n,n-dimethyl-n-octadecyl-, bromide, unii-z5t47r065a, dsab, distearyldimethylammonium bromide, dimethyldioctadecylazanium bromide, distearyl dimethyl ammonium bromide, 1-octadecanaminium, n,n-dimethyl-n-octadecyl-, bromide 1:1
PubChem CID 77293
IUPAC Name dimethyl(dioctadecyl)azanium;bromide
SMILES CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCCCC.[Br-]

Specifications

Melting Point 152°C to 156°C
Odor Odorless
Quantity 1 g
UN Number UN3077
Beilstein 3580323
Formula Weight 630.97
Percent Purity 99%
Chemical Name or Material Dimethyldi-n-octadecylammonium bromide
Hazard Category Danger
Hazard Statement GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
RTECSNumber RG0185984
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.