Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyldioctadecylammonium Bromide 98.0+%, TCI America™
SDP

Catalog No. D1975250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1975250G 250 g
D197525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1975250G Supplier TCI America Supplier No. D1975250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3700-67-2
Molecular Formula C38H80BrN
Molecular Weight (g/mol) 630.969
MDL Number MFCD00041975
InChI Key PSLWZOIUBRXAQW-UHFFFAOYSA-M
Synonym dimethyldioctadecylammonium bromide, n,n-dimethyl-n-octadecyloctadecan-1-aminium bromide, dimethyldistearylammonium bromide, 1-octadecanaminium, n,n-dimethyl-n-octadecyl-, bromide, unii-z5t47r065a, dsab, distearyldimethylammonium bromide, dimethyldioctadecylazanium bromide, distearyl dimethyl ammonium bromide, 1-octadecanaminium, n,n-dimethyl-n-octadecyl-, bromide 1:1
PubChem CID 77293
IUPAC Name dimethyl(dioctadecyl)azanium;bromide
SMILES CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCCCC.[Br-]

Specifications

Melting Point 155°C
Color White
Quantity 250 g
Formula Weight 630.97
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Dimethyldioctadecylammonium Bromide
EINECSNumber (2)-0184&(9)-1971
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.