Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyldiphenylsilane 97.0+%, TCI America™
SDP

Catalog No. D181025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D181025ML 25 mL
D18105ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D181025ML Supplier TCI America Supplier No. D181025ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 778-24-5
Molecular Formula C14H16Si
Molecular Weight (g/mol) 212.367
MDL Number MFCD00041326
InChI Key WJKVFIFBAASZJX-UHFFFAOYSA-N
PubChem CID 136618
IUPAC Name dimethyl(diphenyl)silane
SMILES C[Si](C)(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Color Colorless
Boiling Point 118°C
Quantity 25 mL
Formula Weight 212.37
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Dimethyldiphenylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.