Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl 5-hydroxyisophthalate, 99%, Acros Organics

Click to view available options
Quantity:
10g
25g
Packaging:
Glass bottle
13036-02-7
C10H10O5
210.19
MFCD00134367
DOSDTCPDBPRFHQ-UHFFFAOYSA-N
dimethyl 5-hydroxyisophthalate,5-hydroxyisophthalic acid dimethyl ester,1,3-dimethyl 5-hydroxybenzene-1,3-dicarboxylate,1,3-benzenedicarboxylic acid, 5-hydroxy-, dimethyl ester,dimethyl5-hydroxyisophthalate,timtec-bb sbb000491,5-hydroxy-dimethylisophthalate,1,3-benzenedicarboxylic acid, 5-hydroxy-, 1,3-dimethyl ester,dimethyl-5-hydroxy-benzene-1,3-dicarbonate,methyl 5-hydroxy-3-methoxycarbonyl benzoate
83065
dimethyl 5-hydroxybenzene-1,3-dicarboxylate
COC(=O)C1=CC(=CC(=C1)O)C(=O)OC
13036-02-7
C10H10O5
210.19
MFCD00134367
DOSDTCPDBPRFHQ-UHFFFAOYSA-N
dimethyl 5-hydroxyisophthalate,5-hydroxyisophthalic acid dimethyl ester,1,3-dimethyl 5-hydroxybenzene-1,3-dicarboxylate,1,3-benzenedicarboxylic acid, 5-hydroxy-, dimethyl ester,dimethyl5-hydroxyisophthalate,timtec-bb sbb000491,5-hydroxy-dimethylisophthalate,1,3-benzenedicarboxylic acid, 5-hydroxy-, 1,3-dimethyl ester,dimethyl-5-hydroxy-benzene-1,3-dicarbonate,methyl 5-hydroxy-3-methoxycarbonyl benzoate
83065
dimethyl 5-hydroxybenzene-1,3-dicarboxylate
COC(=O)C1=CC(=CC(=C1)O)C(=O)OC
Color White to Beige
Melting Point 162°C to 165°C
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 99%
Formula Weight 210.19
Physical Form Powder and/or Lumps
Percent Purity ≥98.5%
Chemical Name or Material Dimethyl 5-hydroxyisophthalate, 99%

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.