Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylglyoxime RS (1% w/v) Alcoholic Solution for Nickel Determination, Reagents

Catalog No. 01335418
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-418 1 L
01-335-419 120 mL
01-335-417 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-335-418 Supplier Reagents Holdings Llc Supplier No. CD2610001A
Add to Cart
Add to Cart

Dimethylglyoxime RS (1% w/v) Alcoholic Solution for Nickel Determination, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Dimethylglyoxime RS (1% w/v) Alcoholic Solution for Nickel Determination
CAS 64-17-5, 67-56-1, 95-45-4
Color Colorless
Boiling Point 72°C
Packaging Poly Bottle
Quantity 1 L
Specific Gravity 0.8
Synonym 2,3-Butanedione Dioxime
Percent Purity 1%
Grade Reagent
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.