Learn More
AdipoGen Dimethylindodicarbocyanine
SDP
Supplier:  AdipoGen CDXH0295G005
Encompass_Preferred
Chemical. CAS 54268-70-1. Formula C27H31ClN2. MW 419.01. Cyanine dye. Spectral data Absorption lambdamax 638nm MeOH. Used for the determination of different copper ions CuI, CuII,and CuIII in semiconductor samples and in an environmental friendly Cu extraction procedures. Used as a marker for thermosensitive in situ gelling delivery systems. A different salt form of the compound and laser dye Hexacyanine 2 iodide salt.
Catalog No. 50-256-0693
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.