Learn More
Enzo Life Sciences Dimethyloxaloylglycine (50mg). CAS: 89464-63-1
SDP
Supplier:  Enzo Life Sciences BMLEI3470050
Encompass_Preferred
DMOG is a cell permeable prolyl-4-hydroxylase inhibitor which upregulates HIF activity. HIF activation stimulates angiogenesis in several different models. DMOG also inhibits FIH (Factor Inhibiting HIF), an asparaginyl hydroxylase, which enhances the HIF response. It is active in vivo and attenuates myocardial injury in a rabbit ischemia reperfusion model (20mg/kg). Is expected to act pro-angiogenic. Alternative name: DMOG, N-(Methoxyoxoacetyl)-glycine methyl ester. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (>25mg/ml), 100% ethanol (>25mg/ml) or dimethyl formamide; soluble also in PBS, pH7.2 (10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0734
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.