Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyloxaloylglycine, 98%

Catalog No. AAJ67077MF
Click to view available options
Quantity:
10 mg
50 mg
100 mg

CAS: 89464-63-1 | C6H9NO5 | 175.14 g/mol

DMOG is a cell permeable prolyl-4-hydroxylase inhibitor that regulates hypoxia-inducible factor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89464-63-1
Molecular Formula C6H9NO5
Molecular Weight (g/mol) 175.14
MDL Number MFCD05865098
InChI Key BNJOZDZCRHCODO-UHFFFAOYSA-N
Synonym dmog, dimethyloxalylglycine, dimethyloxaloylglycine, dimethyloxallyl glycine, methyl n-methoxy oxo acetyl glycinate, methyl 2-2-methoxy-2-oxoacetamido acetate, methyl 2-2-methoxy-2-oxoethyl amino-2-oxoacetate, glycine, n-methoxyoxoacetyl-, methyl ester, n-2-methoxy-2-oxoacetyl glycine methyl ester, glycine,n-2-methoxy-2-oxoacetyl-, methyl ester
PubChem CID 560326
IUPAC Name methyl 2-(2-methoxy-2-oxoacetamido)acetate
SMILES COC(=O)CNC(=O)C(=O)OC

Specifications

Color White
Solubility Information Soluble in water at 30mg/ml.
Quantity 50 mg
Formula Weight 175.14
Percent Purity 98%
Physical Form Solid
Chemical Name or Material Dimethyloxaloylglycine
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.