Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylphenylsilane 97.0+%, TCI America™
SDP

Catalog No. D219625ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D219625ML 25 mL
D21965ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D219625ML Supplier TCI America Supplier No. D219625ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 766-77-8
Molecular Formula C8H11Si
Molecular Weight (g/mol) 135.261
MDL Number MFCD00008257
InChI Key OIKHZBFJHONJJB-UHFFFAOYSA-N
Synonym dimethylphenylsilane, dimethyl phenyl silane, phenyldimethylsilane, silane, dimethylphenyl, benzene, dimethylsilyl, dimethylphenyl silane, c8h12si, dimethyl phenyl silyl, dimethyl-phenylsilicon
PubChem CID 6327656
IUPAC Name dimethyl(phenyl)silicon
SMILES C[Si](C)C1=CC=CC=C1

Specifications

Boiling Point 158°C
Color Yellow
Physical Form Liquid
Quantity 25 mL
UN Number 1993
Formula Weight 136.27
Percent Purity ≥97.0% (GC)
Chemical Name or Material Dimethylphenylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.