Learn More
Sigma Aldrich Fine Chemicals Biosciences DIMETHYLPOLYSILOXANE, 100G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences DMPS2C100G
Encompass
Sigma Aldrich | Dimethylpolysiloxane viscosity 200 cSt (25 °C)(lit.) | 100g | DMPS2C-100G | 9016-00-6 | MFCD00084411 | MW: ~9,430Synonym(s):Polydimethylsiloxane
Catalog No. NC2213344
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.