Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimidium bromide, ∽95%, MP Biomedicals™

Catalog No. ICN15095890 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15095890 500 mg
ICN15095891 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15095890 Supplier MP Biomedicals, Inc Supplier No. 0215095890
Add to Cart
Add to Cart

Chemical Identifiers

CAS 518-67-2
Molecular Formula C20H18BrN3
Molecular Weight (g/mol) 380.29
MDL Number MFCD00011757
InChI Key MQOKYEROIFEEBH-UHFFFAOYSA-N
Synonym dimidium bromide, trypadine, 3,8-diamino-5-methyl-6-phenylphenanthridinium bromide, phenanthridinium 1553, phenanthridinium compound 1553, phenanthridinium, 3,8-diamino-5-methyl-6-phenyl-, bromide, 2,7-diamino-10-methyl-9-phenylphenanthridinium bromide, 3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide, phenanthridium 1553
PubChem CID 68207
IUPAC Name 3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide
SMILES [Br-].C[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12

Specifications

Color Black
Assay Percent Range ∽95%
Quantity 500 mg
Formula Weight 380.29
Physical Form Powder
Chemical Name or Material Dimidium bromide
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.