Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DIMT1L Mouse anti-Human, Clone: OTI6E7, liquid, TrueMAB™
SDP

Catalog No. 501683212
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3212 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3212 Supplier OriGene Supplier No. TA810135
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DIMT1 specifically dimethylates two adjacent adenosines in the loop of a conserved hairpin near the 3′-end of 18S rRNA in the 40S particle.

Specifications

Antigen DIMT1L
Applications Western Blot
Classification Monoclonal
Clone OTI6E7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene DIMT1
Gene Accession No. Q9UNQ2
Gene Alias DIM1, DIMT1L, HSA9761, HUSSY5
Gene Symbols DIMT1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-238 of human DIMT1L produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27292
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.