Learn More
Omega Engineering, Inc. DIN RAIL DC CURR SIGNAL SPLTR
Supplier:  Omega Engineering, Inc. DRSPI
Encompass
DIN Rail DC Current Signal Splitters w/ Dual Output.Dimensions: 45 W x 117 H x 122 mm D (1.78 x 4.62 x 4.81"); height includes connectors. --One 4 to 20 mA Input to Two 4 to 20 mA Outputs with Full Isolation. Zero and Span Adjustments for Each Output. Full 1200 V Input/Output/ Power Isolation. Input/Output Loop Status LEDs
Catalog No. NC2330820
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.