Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Dinaline, Antineoplastic Agent, Solid, >98% Purity, 10 mg

Catalog No. 502705627
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5627 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5627 Supplier Abcam Supplier No. AB14456110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Antineoplastic agent with antiproliferative and antitumor effects in vivo.

  • Molecular weight: 227.26 Da
  • Purity: >98%
  • Form: Solid
  • Composition: 100% 4-amino-N-(2-aminophenyl)benzamide
  • Activity: Antineoplastic agent; CI-994 precursor (ab142098); induces reversible changes in amino acid transport, protein metabolism, and DNA synthesis; shows antiproliferative and antitumor effects in vivo
  • Storage: Store at +4°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Dinaline
Shipping Condition Ambient
Content And Storage Store at 4°C.
Buffer 100% 4-amino-N-(2-aminophenyl)benzamide
Form Solid
Substrate Properties Chemical Substrate
Quantity 10 mg
For Use With (Application) Antineoplastic research, antitumor research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.