Learn More
Electron Microscopy Sciences Digital Microscope Model AM73915MZTL, Type: USB 3.0. Magnification: 10x - 140x. LED: White (8)., 1/EA
SDP
Supplier:  Electron Microscopy Sciences AM73915MZTL
Encompass_Preferred
This Dino-Lite Edge USB 3.0 is a longer working distance model that offers uncompressed viewing to deliver exceptional sharpness, improved color accuracy and image quality with a magnification range of 10x - 140x. This model is capable of providing 45FPS at 1280 x 960 resolution, allowing use in a wide range of difficult applications. Automatic Magnification Reading (AMR) and Extended Depth of Field (available on Windows) are included. EDOF lets users automatically capture multiple images and different depths, and automatically combine them into one clear image. Along with the built-on polarizer, interchangeable front caps, and rugged metal housing, this model features flexible LED Control (FLC) that enables partial illumination control options to enhance edge contrast and provide intensity adjustment for more versatility. Type: USB 3.0. Magnification: 10x - 140x . LED: White (8) .
Catalog No. 50-209-1146