Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Dinutuximab Chimeric Recombinant Human Monoclonal Antibody
GREENER_CHOICE

Catalog No. PIMA542000
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542000 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542000 Supplier Invitrogen™ Supplier No. MA542000
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Recombinant Monoclonal Antibody

Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.

Dinutuximab is an IgG1 monoclonal human/mouse chimeric antibody against GD2, a disialoganglioside expressed on tumors of neuroectodermal origin, including human neuroblastoma and melanoma, with highly restricted expression on normal tissues. It is composed of the variable heavy- and light-chain regions of the murine anti-GD2 mAb 14. 18 and the constant regions of human IgG1 heavy-chain and kappa light-chain. By binding to GD2, dinutiximab induces antibody-dependent cell-mediated cytotoxicity and complement-dependent cytotoxicity of tumor cells thereby leading to apoptosis and inhibiting proliferation of the tumor. It is indicated, in combination with granulocyte-macrophage colony-stimulating factor (GM-CSF), interleukin-2 (IL-2), and 13-cis-retinoic acid (RA), for the treatment of pediatric patients with high-risk neuroblastoma who achieve at least a partial response to prior first-line multiagent, multimodality therapy. Despite a high clinical response seen after first-line treatment, the complete eradication of neuroblastoma is rarely achieved and the majority of patients with advanced disease suffer a relapse. Current strategies for treatment include immunotherapy with pharmaceuticals such as dinutuximab to target surviving neuroblastoma cells and to prevent relapse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dinutuximab Chimeric
Applications ELISA, SDS-Page
Classification Recombinant Monoclonal
Concentration 1.3 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias MAb-14.18; MOAB Ch14.18
Host Species Human
Immunogen Human ganglioside GD2.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.