Learn More
Biotium DIO (DIOC18(3))
Supplier:  Biotium 60011
Encompass_Preferred
DiO is a carbocyanine membrane dye that has applications similar to those of DiI. However, DiO has green fluorescent emission and the lateral diffusion rate on the membranes is generally slower than that of DiI. DiO and DiI are often used together in dual color studies. Also available is Neuro-DiO, an improved version of DiO, has better solubility in membranes and does not form non-fluorescent aggregates that can slow dye diffusion. Also offered as ready-to-use CellBrite™ Cytoplasmic Membrane Dyes. Properties: Ex/Em (MeOH) = 484/501 nm; e (MeOH) = 150,000; Yellow orange solid soluble in ethanol; Store at 4°C and protect from light, especially in solution; C53H85ClN2O6; MW: 882; [34215-57-1]
Catalog No. 50-196-4716