Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DIO1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143925
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143925 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143925 Supplier Invitrogen™ Supplier No. PA5143925
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: rat liver tissue, rat kidney tissue, human A549 whole cell. ICC/IF: A549 cell. Flow: 293T cell, ANA-1 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

The protein encoded by this gene is a thiol-requiring propylthiouracil-sensitive oxidoreductase. It activates thyroid hormone by converting the prohormone thyroxine (T4) by outer ring deiodination (ORD) to bioactive 3,3',5-triiodothyronine (T3). It also degrades both hormones by inner ring deiodination (IRD). Alternative splicing results in multiple transcript variants encoding different isoforms. Some, but not all, isoforms contain a selenocysteine (Sec) residue encoded by the UGA codon, which normally signals translation termination. The 3' UTR of Sec-containing genes have a common stem-loop structure, the sec insertion sequence (SECIS), which is necessary for the recognition of UGA as a Sec codon rather than as a stop signal. Additional transcript variants have been described but are not supported by experimental evidence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DIO1
Applications ELISA, Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene Dio1
Gene Accession No. P24389, P49895, Q61153
Gene Alias 5DI; D1; deiodinase, iodothyronine type I; deiodinase, iodothyronine, type I; Dio1; DIOI; hDido1; iodothyronine deiodinase type 1; ITDI1; thyroxine 5'-deiodinase; thyroxine deiodinase; thyroxine deiodinase type I (selenoprotein); thyroxine deiodinase, type I; TXDI1; Type 1 DI; type I iodothyronine deiodinase; Type-I 5'-deiodinase
Gene Symbols Dio1
Host Species Rabbit
Immunogen E.coli-derived human DIO1 recombinant protein (Position: K137-S249).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13370, 1733, 25430
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.