Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dioctyl Phthalate, 98%, Spectrum™ Chemical
SDP

Catalog No. 18610256 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-610-256 20 L Steel Can
18-610-259 500 mL Steel Can
18-610-258 4 L Steel Can
18-610-257 200 L Steel Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-610-256 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D104920LTMT
Add to Cart
Add to Cart

Chemical Identifiers

CAS 117-81-7

Specifications

CAS Max % 1
Boiling Point 384°C
Packaging Steel Can
Quantity 20 L
Percent Purity 0.98
Grade Reagent
Physical Form Liquid
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.