Learn More
Sigma Organic Chemistry Dioctyl sebacate | 1G | 2432-87-3 | MFCD00059269
Supplier: Sigma Organic Chemistry CDS0005591G

Dioctyl sebacate , 1G
About this Item:
CAS #: 2432-87-3
MDL #: MFCD00059269
Molecular Weight: 426.67
UNSPSC Code: 12352200
Chemical Formula: C26H50O4
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.