Learn More
Sigma Organic Chemistry Dioctyl terephthalate | 20L | 6422-86-2 | MFCD00072256 | >=96%
Supplier: Sigma Organic Chemistry 52518920L

Dioctyl terephthalate, 20L
About This Item:
Synonym(s): Bis(2-ethylhexyl) terephthalate
Linear Formula: C6H4-1,4-[CO2CH2CH(C2H5)(CH2)3CH3]2
Molecular Weight: 390.56
CAS Number: 6422-86-2
MDL Number: MFCD00072256
UNSPSC Code: 12162002
Purity: >=96%
Boiling Point: 400 C (lit.)
Melting Point: -48 C
Density: 0.986 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.49 (lit.)
Storage: room temp
EINECS Number: 229-176-9
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.