Learn More
Thermo Scientific Cation Suppressor 2mm [25 EA]
Supplier:  Thermo Scientific 088670CMD
Encompass
2MM internal diameter ion chromatography cation exchange column with sulfonated divinylbenzene-styrene resin for high efficiency separation & analysis of alkali, alkaline earth, transition metals, & amines using suppressed conductivity detection, optimized for isocratic & gradient elution analysis of cationic species in water quality, food, pharmaceutical, & environmental samples.
Catalog No. NC2087525
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.