Learn More
Thermo Scientific™ Dionex™ ASE™ 150/350 Stainless Steel Extraction Cell Bodies and End Caps
Catalog No. 501345454
Encompass
Change view
Click to view available options
Description:
Extraction Cell Body; 10 mL
Extraction Cell End Caps with Frits and Seals
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
50-134-5454 Extraction Cell End Caps with Frits and Seals
50-134-5476 Extraction Cell Body; 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-134-5454 Supplier Thermo Scientific™ Supplier No. 068106
Add to Cart
Add to Cart
Order this long cylindrical portion of the extraction cell if a replacement is needed for your Thermo Scientific™ Dionex™ ASE™ 150 or 350 Accelerated Solvent Extractor.
Choose Thermo Scientific™ Dionex™ ASE™ 150/300 Extraction Cell Bodies—the long cylindrical portion of the extraction cell—if a replacement is needed for your Thermo Scientific™ Dionex™ ASE™ 150 and 350 Accelerated Solvent Extractors. Available in 1, 5, 10, 22, 34, 66, and 100mL sizes, these cell bodies can be ordered individually or in a kit of six.
Please note that these are cell bodies only and do not include the caps and O-ring that screw onto each end of the cell body.
About Dionex ASE Accelerated Solvent Extractor Systems
Dionex ASE systems automate extraction, filtration, and clean-up of solid and semi-solid sample matrices using common solvents at elevated temperatures and pressures. Low- and high-throughput systems are available for a wide variety of applications. ASE systems enable extraction of sample sizes 1 to 100g in minutes and perform unattended extraction of up to 24 samples yet use 50 to 90% less solvent compared to other methods. The chemically inert pathway supports acidic and alkaline matrices and solvents. These flexible, easy-to-operate, cost-effective systems have been approved for use by the U.S. EPA and CLP Program. There are a variety of products that can be added to the Dionex ASE extraction cell to remove moisture, disperse the sample matrix and neutralize mineral acids.
TRUSTED_SUSTAINABILITY
Specifications
Type Extraction Cell End Caps with Frits and Seals
For Use With (Equipment) Dionex ASE 150 or 350 System Stainless Steel Extraction Cell Bodies
Description Extraction Cell End Caps with Frits and Seals
Material Stainless Steel
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.