Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-DIOP, 98%, Thermo Scientific™

Catalog No. AAB2101301
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2101301 0.25 g
AAB2101303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2101301 Supplier Thermo Scientific Chemicals Supplier No. B2101301

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as a ligand for the in situ preparation of chiral hydrogenation catalysts. It also finds it application for the in situ preparation of chiral hydrogenation catalysts, ligand for Rh(I)-catalyzed asymmetric hydrogenation of enamides, 1,4-disilylation of α,β-unsaturated ketones, Heck reaction of alkenyl iodides, and hydroformylation of olefins.

Solubility
Sparingly soluble in water at 25°C 2.1E-5 g/L.

Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37002-48-5
Molecular Formula C31H32O2P2
Molecular Weight (g/mol) 498.543
MDL Number MFCD00009760
InChI Key VCHDBLPQYJAQSQ-LOYHVIPDSA-N
Synonym +-diop, unii-5p52k0ij7t, 4s,5s-2,2-dimethyl-1,3-dioxolane-4,5-diyl bis methylene bis diphenylphosphine, s,s-diop, 4s,5s-+-4,5-bis diphenylphosphinomethyl-2,2-dimethyl-1,3-dioxolane, 2s,3s-+-1,4-bis diphenylphosphino-2,3-o-isopropylidene-2,3-butanediol, 4s,5s-+-bis diphenylphosphinomethyl-2,2-dimethyl-1,3-dioxolane, 4s,5s-4,5-bis diphenylphosphinomethyl-2,2-dimethyl-1,3-dioxolane, +-1,4-bis diphenylphosphino-1,4-dideoxy-2,3-o-isopropylidene-d-threitol, 2s,3s-+-2,3-o-isopropylidene-2,3-dihydroxy-1,4-bis diphenylphosphino butane
PubChem CID 3084647
IUPAC Name [(4S,5S)-5-(diphenylphosphanylmethyl)-2,2-dimethyl-1,3-dioxolan-4-yl]methyl-diphenylphosphane
SMILES CC1(OC(C(O1)CP(C2=CC=CC=C2)C3=CC=CC=C3)CP(C4=CC=CC=C4)C5=CC=CC=C5)C

Specifications

Melting Point 88°C to 90°C
Quantity 0.25 g
Beilstein 1630280
Sensitivity Air sensitive
Solubility Information Sparingly soluble in water at 25°C 2.1E-5g/L.
Optical Rotation +12.5° (c=4.6 in Benzene)
Formula Weight 498.55
Percent Purity 98%
Chemical Name or Material (+)-DIOP
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 253-307-9
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.