Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dioxane ACS, Ricca Chemical
SDP

Catalog No. RSOD01004C Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RSOD01004C 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RSOD01004C Supplier Ricca Chemical Company Supplier No. RSOD01004C
Add to Cart
Add to Cart

Chemical Identifiers

CAS 123-91-1
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

CAS Max % 100
Appearance Clear, Colorless
Quantity 4 L
Packaging Amber Glass Bottle
Residue after Evaporation max. 0.005%
Titratable Acid max. 0.0016%
Water max. 0.05%
Boiling Point 101°C
Percent Purity min. 99%
Grade ACS
Chemical Name or Material Dioxane
Show More Show Less
ShelfLife 36 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.