Learn More
Dioxin in Water, WS Proficiency Testing Material, MilliporeSigma™ Supelco™
Catalog No. 111017018 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mL
Catalog No. Quantity
11-101-7018 2 mL
1 options
Catalog No. 11-101-7018 Supplier MilliporeSigma Supelco Supplier No. PE13732ML
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
This concentrate contains 2,3,7,8-TCDD in toluene.Sample preparationPour 1000-mL of reagent grade DI water into a extraction vessel. Open ampule and transfer 1 mL of concentrate to the extraction vessel using Class A glassware. Analyze as required by your normal procedures. For calculation purposes assume 1 liter sample volume.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Ampule
Quantity 2 mL
Concentration or Composition (by Analyte or Components) 2,3,7,8-TCDD 20 - 100 pg/L