Learn More
Dioxins and Furans in Water by 8280, PT Proficiency Testing Material, MilliporeSigma™ Supelco™
Catalog No. 111017014 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mL
Catalog No. Quantity
11-101-7014 2 mL
1 options
Catalog No. 11-101-7014 Supplier MilliporeSigma Supelco Supplier No. PE11022ML
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
This concentrate contains up to 16 dioxins and furans in toluene.Sample preparationPour 1000-mL of reagent grade DI water into a extraction vessel. Open ampule and transfer 1 mL of concentrate to the extraction vessel using Class A glassware. Analyze as required by your normal procedures. Assume a 1-Liter sample volume for calculation purposes.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Ampule
Quantity 2 mL
Concentration or Composition (by Analyte or Components) 1,2,3,4,6,7,8-Hpcdf 1-2000 ng/L; 1,2,3,4,7,8,9-Hpcdf 1-2000 ng/L; 1,2,3,4,6,7,8-Hpcdd 1-2000 ng/L; Total Heptachlorodibenzo-p-dioxin (Total HPCDD) 1-2000 ng/L; Total Heptachlorodibenzofuran (Total HPCDF) 1-2000 ng/L; 1,2,3,4,7,8-Hxcdd 1-2000 ng/L; 1,2,3,6,7,8-Hxcdd 1-2000 ng/L; 1,2,3,7,8,9-Hxcdd 1-2000 ng/L; Hxcdd, total 1-2000 ng/L; 1,2,3,4,7,8-Hxcdf 1-2000 ng/L; 1,2,3,6,7,8-Hxcdf 1-2000 ng/L; 1,2,3,7,8,9-Hxcdf 1-2000 ng/L; 2,3,4,6,7,8-Hxcdf 1-2000 ng/L; Total Hexachlorodibenzofuran (Total HxCDF) 1-2000 ng/L; 1,2,3,4,6,7,8,9-OCDF 1-2000 ng/L; 1,2,3,4,6,7,8,9-OCDD 1-2000 ng/L; 1,2,3,7,8-Pecdd 1-2000 ng/L; 1,2,3,7,8-Pecdf 1-2000 ng/L; 2,3,4,7,8-Pecdf 1-2000 ng/L; Pecdf, total 1-2000 ng/L; Pecdd, total 1-2000 ng/L; TCDD, total 1-2000 ng/L; 2,3,7,8-TCDF 1-2000 ng/L; TCDF, total 1-2000 ng/L; 2,3,7,8-TCDD 1-2000 ng/L; PCDF, total 1-2000 ng/L; PCDD, total 1-2000 ng/L; Total Polychlorinated dibenzo-p-dioxins and total Polychlorinated dibenzofurans (Total PCDD/Fs) 1-2000 ng/L