Learn More
Dioxins in Water, WP Proficiency Testing Material, MilliporeSigma™ Supelco™
Catalog No. 111017013 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mL
Catalog No. Quantity
11-101-7013 2 mL
1 options
Catalog No. 11-101-7013 Supplier MilliporeSigma Supelco Supplier No. PE12952ML
This proficiency testing sample is produced in accordance with ISO/IEC 17043:2010. All information regarding the use of this material can be found in the reporting packet supplied for each sample.
This concentrate contains up to 16 dioxins and furans in tolueneSample preparationPour 1000-mL of reagent grade DI water into a extraction vessel. Open ampule and transfer 1 mL of concentrate to the extraction vessel using Class A glassware. Analyze as required by your normal procedures. Assume a 1-Liter sample volume for calculation purposes.
TRUSTED_SUSTAINABILITY
Specifications
Packaging Ampule
Quantity 2 mL
Concentration or Composition (by Analyte or Components) 1,2,3,4,6,7,8-Hpcdf 50 - 2000 pg/L; 1,2,3,4,7,8,9-Hpcdf 50 - 2000 pg/L; 1,2,3,4,6,7,8-Hpcdd 50 - 2000 pg/L; Total Heptachlorodibenzo-p-dioxin (Total HPCDD) 50 - 4000 pg/L; Total Heptachlorodibenzofuran (Total HPCDF) 50 - 4000 pg/L; 1,2,3,4,7,8-Hxcdd 50 - 2000 pg/L; 1,2,3,6,7,8-Hxcdd 50 - 2000 pg/L; 1,2,3,7,8,9-Hxcdd 50 - 2000 pg/L; Hxcdd, total 50 - 6000 pg/L; 1,2,3,4,7,8-Hxcdf 50 - 2000 pg/L; 1,2,3,6,7,8-Hxcdf 50 - 2000 pg/L; 1,2,3,7,8,9-Hxcdf 50 - 2000 pg/L; 2,3,4,6,7,8-Hxcdf 50 - 2000 pg/L; Total Hexachlorodibenzofuran (Total HxCDF) 50 - 8000 pg/L; 1,2,3,4,6,7,8,9-OCDF 50 - 2000 pg/L; 1,2,3,4,6,7,8,9-OCDD 50 - 2000 pg/L; 1,2,3,7,8-Pecdd 50 - 2000 pg/L; 1,2,3,7,8-Pecdf 50 - 2000 pg/L; 2,3,4,7,8-Pecdf 50 - 2000 pg/L; Pecdf, total 50 - 6000 pg/L; Pecdd, total 50 - 2000 pg/L; TCDD, total 50 - 10000 pg/L; 2,3,7,8-TCDF 50 - 2000 pg/L; TCDF, total 50 - 10000 pg/L; 2,3,7,8-TCDD 50 - 2000 pg/L; PCDF, total 50 - 10000 pg/L; PCDD, total 50 - 10000 pg/L; Total Polychlorinated dibenzo-p-dioxins and total Polychlorinated dibenzofurans (Total PCDD/Fs) 1 50 - 20000 pg/L