Learn More
ENG Scientific Dip Stain Kits (Wright and Wright Giemsa)
Description
- Solutions made from certified dyes
- Solutions should be replenished during staining period
- During cold weather, allow solutions to return to room temperature and shake well before use
- Dip Stain Kit includes: Wright or Wright Giemsa Dip Stain, methanol, buffers, stabilizers
- Dip Stain Kit Replacement Solutions include: 1 gallon of solution
- For in-vitro diagnostic use only
Specifications
Specifications
| Quantity | 4 x 950 mL |
| Storage Requirements | Room Temperature |
| For Use With (Application) | For in-vitro diagnostic use only |
| Packaging | Complete Kit (1 each Solutions I - IV) |
| Product Type | Wright Dip Stain Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.