Learn More
Abcam Dipeptidyl Peptidase IV Inhibitor Screening Assay Kit, Fluorometric, 96 Test
Description
- Assay type: Enzyme activity
- Detection method: Fluorescent
- Purpose: Assess DPP4 inhibitors
- Readout: Fluorometric plate reader
- Citations: 7
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| No. of Reactions | 96 Tests |
| Substrate Properties | Peptide-based Substrate |
| Target Molecule | Dipeptidyl Peptidase IV |
| Product Type | Western Blot Kit |
| For Use With (Equipment) | Fluorometric Plate Reader |
| Format | Kit |
| Detection Method | Fluorescence |
| Quantity | 96 Tests |
| For Use With (Application) | Inhibitor screening |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.