Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenhydramine 98.0+%, TCI America™
SDP

Catalog No. D474425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D474425G 25 g
D47445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D474425G Supplier TCI America Supplier No. D474425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 58-73-1
Molecular Formula C17H21NO
Molecular Weight (g/mol) 255.361
MDL Number MFCD00274173
InChI Key ZZVUWRFHKOJYTH-UHFFFAOYSA-N
Synonym 2-Diphenylmethoxy-N,N-dimethylethylamine, 2-(Benzhydryloxy)-N,N-dimethylethylamine
PubChem CID 3100
ChEBI CHEBI:4636
IUPAC Name 2-benzhydryloxy-N,N-dimethylethanamine
SMILES CN(C)CCOC(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Color Yellow
Quantity 25 g
Formula Weight 255.36
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Diphenhydramine
EINECSNumber (9)-0484&(9)-0690
RTECSNumber KR6825000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.