Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyl carbonate, 99%

Catalog No. p-4258982
Change view
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC117231000 100 g Plastic bottle
AC117230010 1 kg Plastic bottle
AC117230025 2.5 kg Plastic bottle
AC117230100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC117231000 Supplier Thermo Scientific Chemicals Supplier No. 117231000
Only null left
Add to Cart

CAS: 102-09-0 | C13H10O3 | 214.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-09-0
Molecular Formula C13H10O3
Molecular Weight (g/mol) 214.22
InChI Key ROORDVPLFPIABK-UHFFFAOYSA-N
Synonym carbonic acid, diphenyl ester, phenyl carbonate, diphenylcarbonate, carbonic acid diphenyl ester, phenyl carbonate pho 2co, unii-ywv401idyn, ph2co3, pho 2co, ywv401idyn, phenyl phenoxyformate
PubChem CID 7597
ChEBI CHEBI:34722
IUPAC Name diphenyl carbonate
SMILES C1=CC=C(C=C1)OC(=O)OC2=CC=CC=C2

Specifications

Melting Point 78.0°C to 81.0°C
Color White
Boiling Point 301.0°C to 302.0°C
Flash Point 168°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula (C6H5O)2CO
Quantity 100 g
Beilstein 06,158
Merck Index 15,3354
Solubility Information Solubility in water: insoluble. Other solubilities: solulbe in acetone, hot alcohol, benzene, carbon, tetrachloride, ether, glacial acetic acid and, other organic solvents
Formula Weight 214.22
Percent Purity 99%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material Diphenyl carbonate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Avoid release to the environment.
EINECSNumber 203-005-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.