Learn More
Sigma Organic Chemistry Diphenyl ether | 3KG | 101-84-8 | MFCD00003034
Supplier: Sigma Organic Chemistry P241013KG

Diphenyl ether, 3KG
About This Item:
Synonym(s): Diphenyl oxide; Phenyl ether
Linear Formula: (C6H5)2O
Molecular Weight: 170.21
CAS Number: 101-84-8
MDL Number: MFCD00003034
UNSPSC Code: 12352100
Purity: 0.99
Boiling Point: 259 C (lit.)
Melting Point: 25-27 C (lit.)
Density: 1.073 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.579 (lit.)
Storage: room temp
EINECS Number: 202-981-2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.