Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diphenyl-n-propylphosphine, 97%

Catalog No. AAL0240203
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0240203 1 g
AAL0240206 5 g
AAL0240214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0240203 Supplier Thermo Scientific Chemicals Supplier No. L0240203
Add to Cart

CAS: 7650-84-2 | C15H17P | 228.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7650-84-2
Molecular Formula C15H17P
Molecular Weight (g/mol) 228.28
MDL Number MFCD00015216
InChI Key AAXGWYDSLJUQLN-UHFFFAOYSA-N
Synonym diphenyl-n-propylphosphine, phosphine, diphenylpropyl, propyldiphenylphosphine, n-propyldiphenylphosphine, diphenyl propyl phosphine, diphenylphosphinopropane, diphenyl propyl phosphane, diphenylphospinopropane, di phenyl-propylphosphane
PubChem CID 82100
IUPAC Name diphenyl(propyl)phosphane
SMILES CCCP(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Density 1.008
Boiling Point 163°C to 164°C (14 mmHg)
Flash Point >110°C (230°F)
Odor Unpleasant
Refractive Index 1.6035
Quantity 1 g
Beilstein 642795
Sensitivity Air Sensitive
Formula Weight 228.28
Percent Purity 97%
Chemical Name or Material Diphenyl-n-propylphosphine
Show More Show Less
Hazard Category H302-H315-H319
Hazard Statement GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
EINECSNumber 231-607-0
RTECSNumber SY9150000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.